missing translation for 'onlineSavingsMsg'
Learn More
Beryllium foil, 0.1mm (0.004in) thick, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11388778

Beryllium foil, 0.1mm (0.004in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11388778
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11388778

Brand: Thermo Scientific Alfa Aesar 041643.FF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-41-7
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]

Specifications

Quantity 25 x 25 mm
Percent Purity 99.8%
Odor Odorless
Weight ≈0.12g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Beryllium foil, 0.1mm (0.004 in.) thick, Vacuum tight

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG09W024 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041642.FI,041642.FF,041643.FF
Lot NumberT01A010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041642.FI,041642.FF,041643.FF
Lot NumberX19B030 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041642.FI,041642.FF,041643.FF
Lot NumberH02K02 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041642.FI,041642.FF,041643.FF
Lot NumberT24B065 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041642.FI,041642.FF,041643.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Beryllium foil, 0.1mm (0.004in) thick, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.