missing translation for 'onlineSavingsMsg'
Learn More

Beryllium foil, 0.5mm (0.02in) thick, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11347078
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-41-7
Molecular Formula Be
Molecular Weight (g/mol) 9.01
MDL Number MFCD00134032
InChI Key ATBAMAFKBVZNFJ-UHFFFAOYSA-N
Synonym glucinium, dust, metal, powder, metallic, berilio, beryllium, elemental, glucinum, beryllium-9, and compounds
PubChem CID 5460467
ChEBI CHEBI:33783
IUPAC Name beryllium
SMILES [Be]

Specifications

Quantity 25 x 25 mm
Percent Purity 99.4%
Odor Odorless
Weight ≈0.6g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Beryllium foil, 0.5mm (0.02 in.) thick, Vacuum tight

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'