missing translation for 'onlineSavingsMsg'
Learn More

Beryllium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11353637
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Beryllium oxide is used in rocket engines. It is also used as a structural ceramic for high-performance microwave devices, vacuum tubes, magnetrons, and gas lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-56-9
Molecular Formula BeO
Molecular Weight (g/mol) 25.01
MDL Number MFCD00003457
InChI Key LTPBRCUWZOMYOC-UHFFFAOYSA-N
Synonym beryllium oxide, beryllia, bromellete, thermalox, beryllium monoxide, natural bromellite, thermalox 995, ccris 83, beryllium oxide beryllium and beryllium compounds
PubChem CID 14775
ChEBI CHEBI:62842
IUPAC Name oxoberyllium
SMILES [Be]=O

Specifications

Boiling Point 4300°C
Melting Point 2575°C
Physical Form Powder
Quantity 1 g
UN Number UN1566
Merck Index 14,1172
Formula Weight 25.01
Percent Purity 99.95%
Odor Odorless
Density 3.01g/mL
Assay (metals basis)
Chemical Name or Material Beryllium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'