missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beryllium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Bottle
Specifications
Specifications
Concentration | 10mg/L |
Packaging | Bottle |
Quantity | 50 mL |
Solvent or Matrix | HNO3 |
Chemical Name or Material | Beryllium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction