missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beryllium sulfate tetrahydrate, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
In Industries it is used for beryllium salt making, ceramics and chemical reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Lump |
Quantity | 25 g |
UN Number | UN1566 |
Merck Index | 14,1175 |
Solubility Information | Freely soluble in water; Insoluble in alcohol |
Formula Weight | 177.14 (105.07 Anhydrous) |
Percent Purity | 99.99% |
Density | 1.1713 g/mL |
Assay | (metals basis) |
Chemical Name or Material | Beryllium sulfate tetrahydrate |
Safety and Handling
Product Identifier
- Beryllium sulfate tetrahydrate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Beryllium sulfate tetrahydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'