missing translation for 'onlineSavingsMsg'
Learn More

beta Amyloid Mouse anti-Human, Porcine, Clone: 5C2A1, Proteintech

Mouse Monoclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen beta Amyloid
Clone 5C2A1
Concentration 1.12 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Productos 2
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
Código de producto Marca Quantity Precio Cantidad y disponibilidad  
16847455
missing translation for 'viewDocuments'
Proteintech
60342-1-IG-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16837455
missing translation for 'viewDocuments'
Proteintech
60342-1-IG-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

Amyloid beta peptide (Abeta) is the major constituent of amyloid plaques in the brains of individuals afflicted with Alzheimer′s disease. This peptide is generated from the beta-amyloid precursor protein (beta APP) in a two-step process. The first step involves cleavage of the extracellular, amino-terminal domain of beta APP. Protein cleavage is performed by an aspartyl protease termed beta-secretase (BACE). This enzyme is synthesized as a propeptide that must be modified to the mature and active form by the prohormone convertase, furin. Beta APP cleavage by the mature form of BACE results in the cellular secretion of a segment of beta APP and a membrane-bound remt. This remt is then processed by another protease termed gamma-secretase. Gamma-secretase cleaves an intra-membrane site in the carboxyl-terminal domain of beta APP, thus generating the amyloid beta peptide. Gamma-secretase is believed to be a multi-subunit complex containing presenilin-1 and 2 as central components. Found associated with the presenilins is the transmembrane glycoprotein nicastrin. Nicastrin has been found to bind to the carboxyl-terminus of betaAPP and helps to modulate the production of the amyloid beta peptide Also found in the neurofibrillary lesions in Alzheimer′s disease is the protein termed Tau. Tau is a neuronal microtubule-associated protein found predomitly on axons. The function of tau is to promote tubulin polymerization and stabilize microtubules. Tau, in its hyperphosphorylated form, is the major component of paired helical filaments (PHF), which is the building block of neurofibrillary lesions in Alzheimer′s disease brain.
Especificaciones

Especificaciones

beta Amyloid
1.12 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A4, AAA, ABETA, ABPP, AD1, AICD 50, AICD 57, AICD 59, AID(50), AID(57), AID(59), Amyloid beta A4 protein, amyloid P, APP, APPI, beta Amyloid, CTFgamma, CVAP, Gamma CTF(50) C31, PN II, PN2, PreA4, Protease nexin II, S APP alpha Soluble APP beta
APP
IgG2a
Protein A
Antibody
5C2A1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Porcine
P05067, P79307
351, 397663
beta Amyloid Fusion Protein Ag0769
Primary
-20°C
APP
Videos
SDS
Documentos

Documentos

Certificados
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado