Learn More
Fisherbrand™ Radiation Safety Storage Boxes
Radiation safety boxes that are ideal for protection against Beta and Gamma radiation.
Brand:  Fisherbrand™ CSR-B53
Product Code. 12317127
  • 1015.00 € / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Beta – 53L – (40 x 49 x 27cm), use with bag CSR-BAG5
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
For storing radioactive materials, To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
53 L
400mm
Radiation Storage Box