Learn More
beta-Bromophenetole, 98%, Thermo Scientific™
23.50 € - 145.00 €
Chemical Identifiers
CAS | 589-10-6 |
---|---|
Molecular Formula | C8H9BrO |
Molecular Weight (g/mol) | 201.06 |
MDL Number | MFCD00000234 |
InChI Key | JJFOBACUIRKUPN-UHFFFAOYSA-N |
Synonym | 2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene |
PubChem CID | 68526 |
IUPAC Name | 2-bromoethoxybenzene |
SMILES | C1=CC=C(C=C1)OCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10560512
|
Thermo Scientific Acros
107250050 |
5 g | Glass bottle |
23.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10325980
|
Thermo Scientific
107251000 |
100 g | Glass bottle | ||||||
Description
Chemical Identifiers
589-10-6 | |
201.06 | |
JJFOBACUIRKUPN-UHFFFAOYSA-N | |
68526 | |
C1=CC=C(C=C1)OCCBr |
C8H9BrO | |
MFCD00000234 | |
2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene | |
2-bromoethoxybenzene |
Specifications
589-10-6 | |
100.0 | |
Colorless | |
65°C | |
97.5% min. (GC) | |
C6H5OCH2CH2Br | |
MFCD00000234 | |
2-bromoethoxy benzene, 2-bromoethyl phenyl ether, beta-bromophenetole, 2-phenoxyethylbromide, 2-phenoxyethyl bromide, 1-bromo-2-phenoxyethane, benzene, 2-bromoethoxy, beta-phenoxyethyl bromide, 2-bromoethylphenylether, 1-2-bromoethoxy benzene | |
C1=CC=C(C=C1)OCCBr | |
201.06 | |
201.06 | |
Liquid After Melting |
97.5 | |
33.5°C to 34.0°C | |
144.0°C (40.0 mmHg) | |
Authentic | |
C8H9BrO | |
1.5550 to 1.5570 | |
06, 142 | |
JJFOBACUIRKUPN-UHFFFAOYSA-N | |
2-bromoethoxybenzene | |
68526 | |
98% | |
β-Bromophenetole, 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-634-4
missing translation for 'tsca' : TSCA
- ß-Bromophenetole
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only