Learn More
Fisherbrand™ Beta/Gamma Protection Duoshield Box
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™ CSR-SDUO
Product Code. 12327197
  • 3370.00 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent shielding
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
Transparent
beta/gamma radiation protection
Duoshield box
L x W : 45 x 30cm (17.71 x 11.81 in.)
Curved base
Acrylic
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Beta/Gamma Protection Duoshield Box > Beta/Gamma Protection Duoshield Box