missing translation for 'onlineSavingsMsg'
Learn More

beta-Ionone, 96%, Thermo Scientific Chemicals

47.74 € - 150.75 €

Chemical Identifiers

CAS 79-77-6
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00001549
InChI Key PSQYTAPXSHCGMF-BQYQJAHWSA-N
Synonym beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
PubChem CID 638014
ChEBI CHEBI:32325
IUPAC Name (E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
SMILES CC1=C(C(CCC1)(C)C)C=CC(=O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11445413
View Documents
Thermo Scientific Alfa Aesar
A13400.22
100 g
47.74 €
100g
Please to purchase this item. Need a web account? Register with us today!
11455413
View Documents
Thermo Scientific Alfa Aesar
A13400.36
500 g
150.75 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important fragrance chemical used in perfumery. The combination of α-ionone and β-ionone is characteristic of the scent of violets and used with other components in flavoring to recreate their scent.

Solubility
Soluble in water (0.11 mg/mI at 20°C).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

79-77-6
192.302
PSQYTAPXSHCGMF-BQYQJAHWSA-N
638014
(E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
C13H20O
MFCD00001549
beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
CHEBI:32325
CC1=C(C(CCC1)(C)C)C=CC(=O)C

Specifications

79-77-6
0.943
112°C (233°F)
1.52
1909544
beta-ionone, trans-beta-ionone, e-beta-ionone, 4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, .beta.-ionone, 3e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, 4-2,6,6-trimethyl-1-cyclohexenyl-3-buten-2-one, unii-a7nrr1hlh6, e-4-2,6,6-trimethylcyclohex-1-en-1-yl but-3-en-2-one, beta-e-ionone
PSQYTAPXSHCGMF-BQYQJAHWSA-N
(E)-4-(2,6,6-trimethylcyclohexen-1-yl)but-3-en-2-one
638014
192.3
beta-Ionone
-49°C
126°C to 128°C (12 mmHg)
C13H20O
MFCD00001549
14,5056
Soluble in water (0.11mg/mI at 20°C).
CC1=C(C(CCC1)(C)C)C=CC(=O)C
192.302
CHEBI:32325
96%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 201-224-3

missing translation for 'rtecsNumber' : EN0350000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only