Learn More
Fisherbrand™ Beta Mini Radiation Storage Containers
Ideal solution for storing radioactive materials, to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes.
Brand:  Fisherbrand™ CSR-B0.4
Product Code. 12397097
  • 646.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Beta – Mini (5.5 x 8.5 x 8.5cm)
  • For storing radioactive materials, to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
Specifications
Specifications
To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
55mm
Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
Radiation Storage Box
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products