missing translation for 'onlineSavingsMsg'
Learn More

beta-Naphthol Violet, Thermo Scientific™

Catalog Number 11455687
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7143-21-7
C16H9N3Na2O9S2
497.36
MFCD00021457
QMNPQXAVQXMXTD-UHFFFAOYSA-L
44135658
disodium;4-[(4-nitrophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
C1=CC(=CC=C1NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[N+](=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
7143-21-7
C16H9N3Na2O9S2
497.36
MFCD00021457
QMNPQXAVQXMXTD-UHFFFAOYSA-L
44135658
disodium;4-[(4-nitrophenyl)hydrazinylidene]-3-oxonaphthalene-2,7-disulfonate
C1=CC(=CC=C1NN=C2C3=C(C=C(C=C3)S(=O)(=O)[O-])C=C(C2=O)S(=O)(=O)[O-])[N+](=O)[O-].[Na+].[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Beta-Naphthol violet exhibits antiseptic properties and is used in the synthesis of certain azo dyes and antioxidants for rubber. It can also be used in the detection of the ferric ion which turns the Beta-Naphthol violet solution green.

Solubility
Soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Solubility Information Soluble in water.
Formula Weight 497.37
Odor Odorless
Quantity 10 g
Chemical Name or Material beta-Naphthol Violet
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'