missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%, Thermo Scientific Chemicals

Product Code. 15470247
Click to view available options
Quantity:
250 mg
1 g

Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

Solubility
Soluble in water (50 mg/ml), 0.001% aqueous buffer, pH 7.0, methanol, and ethanol (less soluble). Insoluble in ether, and ethyl acetate.

Notes
Protect from light and moisture. Keep cool and dry. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24292-60-2
Molecular Formula C21H26N7Na2O17P3
Molecular Weight (g/mol) 787.37
MDL Number MFCD00065390
InChI Key WSDDJLMGYRLUKR-MWLGWILCNA-L
Synonym nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
PubChem CID 131851653
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
SMILES [Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water (50mg/ml),0.001% aqueous buffer,pH 7.0,methanol,and ethanol (less soluble). Insoluble in ether,and ethyl acetate.
Formula Weight 787.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate disodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'