Learn More
Fisherbrand™ Beta Radiation Safe Hourglass Shield
Provides radiation protection shielding for use in laboratories.
Brand:  Fisherbrand™ CSR-S4
Product Code. 11517253
  • 528.00 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transparent shielding
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Specifications
Acrylic
Flat base
L x B: 45 x 30 cm (17.71 x 11.81 in.)
Beta radiation protection
Hourglass shield