missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Beta Radiation Safe Storage Box for Four Way Racks
Provides radiation protection shielding for use in laboratories.
Brand: Fisherbrand™ CSR-B3.5
This item is not returnable.
View return policy
Description
- Manufactured with hinged lids
- Transparent shielding
- Serves as solution for short-term storage of radioactive waste or radioisotopes
- Constructed of acrylic provides easy fabrication and manufacturability
Specifications
17.5 x 15 x 14cm (6.89 x 5.90 x 5.51 in.) | |
Radioisotopes or radioactive waste | |
Storage Box for four way racks |
Beta radiation shielded | |
Hinged lid | |
Acrylic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Beta Radiation Safe Storage Box for Four Way Racks > Beta Radiation Safe Storage Box with Four Way Racks
Spot an opportunity for improvement?Share a Content Correction