missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Beta Radiation Shields

Ideal for protection from Beta radiation, Fisherbrand™ radiation shields are supplied in small, medium and large sizes and with a 15cm-deep curved base for use with safety trays, or a 30x30cm flat base for under-the-bench protection.

Brand:  Fisherbrand™ CSR-S2

 View more versions of this product

Product Code. 12387177

  • 356.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Beta – Large Fixed 15° Angle – Flat Base 35 x 53cm, Base 35 x 30cm
  • A range of angles offers increased manoeuvrability, while clear optical acrylic aids visualisation
  • Shield, fixed 15 degree angle, flat base, dimensions 530h x 350w x 300d mm 
Specifications

Specifications

Flat Base
15° Angle
Beta Radiation Shield
531 x 350 x 300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.