missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Beta Radiation Storage Mini Box Inserts

Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.

Brand:  Fisherbrand™ CSR-R0.5

Product Code. 12367157

  • 46.80 € / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Cleaver radiation shielding mini insert for 20 x 0.5ml tubes
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Various inserts to fit inside Mini boxes
Specifications

Specifications

To hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
40mm
Radiation Storage Mini Box Insert
FB58417 20 x 1.5mL Tubes
Microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
80mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.