missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betaine, 5M Solution, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
85.80 € - 441.87 €
Chemical Identifiers
CAS | 107-43-7 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.148 |
MDL Number | MFCD00012123 |
InChI Key | KWIUHFFTVRNATP-UHFFFAOYSA-N |
Synonym | betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin |
PubChem CID | 247 |
ChEBI | CHEBI:17750 |
IUPAC Name | 2-(trimethylazaniumyl)acetate |
SMILES | C[N+](C)(C)CC(=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15855428
|
Thermo Scientific
J77507.AB |
10 mL | Plastic bottle |
85.80 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15865428
|
Thermo Scientific
J77507.AE |
100 mL | Plastic bottle |
441.87 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
107-43-7 | |
117.148 | |
KWIUHFFTVRNATP-UHFFFAOYSA-N | |
247 | |
2-(trimethylazaniumyl)acetate |
C5H11NO2 | |
MFCD00012123 | |
betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
CHEBI:17750 | |
C[N+](C)(C)CC(=O)[O-] |
Specifications
107-43-7 | |
MFCD00012123 | |
betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
C[N+](C)(C)CC(=O)[O-] | |
117.148 | |
CHEBI:17750 | |
Solution |
C5H11NO2 | |
3537113 | |
KWIUHFFTVRNATP-UHFFFAOYSA-N | |
2-(trimethylazaniumyl)acetate | |
247 | |
Molecular Biology, Ultrapure | |
Betaine |
Safety and Handling
missing translation for 'einecsNumber' : 203-490-6
missing translation for 'rtecsNumber' : DS5900000
missing translation for 'tsca' : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only