Learn More
Texwipe™ BetaMop™ II
Low contamination properties make this the perfect mop for floor cleaning in critical environments
Brand:  Texwipe™ TX7106
 View more versions of this product
Product Code. 11581214
  • 168.00 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Specifications
BetaMop II Mop
Corrosion
Wring-bucket Systems
Polyester
Autoclavable
Handle and Head Refill
For floor cleaning in both electronic cleanrooms and sterile environments