missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaSil™ Phenyl/Hexyl Guard Cartridges
Drop-in guard cartridges offer convenience, economy and effective protection for extending column lifetime
411.13 €
Specifications
Length (Metric) | 10 mm |
---|---|
Stationary Phase | Phenyl/Hexyl |
Requires: Thermo Scientific UNIGUARD™ direct connection or stand alone holder, sold separately.
Description
- The 10mm design offers maximum protection with minimal increase in retention
- Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
Features of Betasil Phenyl/Hexyl Solid Phase
- Ligand on 100Å silica
- High surface area with high bonded phase coverage
- Betasil Phenyl/Hexyl columns offer unique selectivity
Specifications
10 mm | |
Phenyl/Hexyl |