missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaSil™ Silica Guard Cartridges

Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.

330.48 € - 451.39 €

Specifications

Particle Size 5 μm
Length (Metric) 10 mm
Product Line BetaSil
View More Specs

Products 4
Product Code Brand Diameter (Metric) Length (Metric) Particle Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Particle Size Price Quantity & Availability  
10187644
missing translation for 'viewDocuments'
Thermo Scientific™
70005-012101
2.1 mm 10 mm 5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10279174
missing translation for 'viewDocuments'
Thermo Scientific™
70005-013001
3 mm 10 mm 5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10239224
missing translation for 'viewDocuments'
Thermo Scientific™
70005-014001
4 mm 10 mm 5 μm
451.39 €
Pack of 4
In Stock
to see stock.
Add to basket
 
10514855
missing translation for 'viewDocuments'
Thermo Scientific™
70005-011001
1 mm I.D. 10 mm 5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.

Thermo Scientific™ BETASIL™ Silica HPLC columns are based on a highly pure, 100Å silica designed for small molecule analysis. BETASIL Silica columns are used for normal phase separations. These non-bonded silica columns are a powerful and efficient tool for separating non-polar and moderately polar compounds.

  • 100Å high purity silica
  • Used for the separation of non-polar and moderately polar compounds.
Specifications

Specifications

5 μm
10 mm
BetaSil
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.