missing translation for 'onlineSavingsMsg'
Learn More

Bevantolol hydrochloride, Thermo Scientific™

Product Code. 17372501
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 42864-78-8
Molecular Formula C20H28ClNO4
Molecular Weight (g/mol) 381.90
MDL Number 00941389
InChI Key FJTKCFSPYUMXJB-UHFFFAOYNA-N
IUPAC Name hydrogen 1-{[2-(3,4-dimethoxyphenyl)ethyl]amino}-3-(3-methylphenoxy)propan-2-ol chloride
SMILES [H+].[Cl-].COC1=CC=C(CCNCC(O)COC2=CC=CC(C)=C2)C=C1OC

Specifications

Concentration 97.5% to 102.5%
Packaging Glass Bottle
Quantity 250 mg
Color White to Yellow to Beige
Certificate of Analysis Available
Concentration or Composition Notes Titration Argentometric
Merck Index 1194
Formula Weight 381.9
Melting Point 225°C to 228°C
Solubility Slightly soluble in water; Soluble in methanol and chloroform; Insoluble in ether, ethyl acetate and hexane
Physical Form Powder
Chemical Name or Material Bevantolol hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.