Learn More
Keystone Cleanroom Products BHD Blue Autoclaveable Wrapper
BIOPHARMAGREENER_CHOICE
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an adequate microbial barrier from external sources of contamination.
Brand:  Keystone Cleanroom Products BHDSHEET4040
 View more versions of this product
Product Code. 17738197
  • 1035.00 € / Pack of 250
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for Steam Sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient Microbial Barrier
  • Resists Tearing and Punctures
  • Does not contain any natural latex rubber
  • Processed and Packaged in ISO Certified Cleanrooms
  • Certificates of Conformance sent with shipment
Specifications
Specifications
40 x 40 in.