missing translation for 'onlineSavingsMsg'
Learn More

BI 78D3, Tocris Bioscience™

Product Code. 18065512
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective, competitive JNK inhibitor

Competitive c-Jun N-terminal kinase (JNK) inhibitor (IC50 = 280 nM) that displays > 100 fold selectivity over p38α and no activity at mTOR and PI-3K. Inhibits JNK interacting protein 1 (JIP1)-JNK binding (IC50 = 500 nM) and prevents JNK substrate phosphorylation. Blocks JNK-dependent Con A-induced liver damage and restores insulin sensitivity in a mouse model of type II diabetes.

Chemical Identifiers

CAS 883065-90-5
Molecular Formula C13H9N5O5S2
Molecular Weight (g/mol) 379.365
InChI Key QFRLDZGQEZCCJZ-UHFFFAOYSA-N
Synonym 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-3-5-nitrothiazol-2-yl thio-1h-1,2,4-triazol-5 4h-one, 4-2,3-dihydro-1,4-benzodioxin-6-yl-2,4-dihydro-5-5-nitro-2-thiazolyl thio-3h-1,2,4-triazol-3-one, 4-2,3-dihydro-1,4-benzodioxin-6-yl-3-5-nitro-1,3-thiazol-2-yl sulfanyl-1h-1,2,4-triazol-5-one, 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-3-5-nitro-thiazol-2-yl thio-1h-1,2,4-triazol-5 4h-one, 4-2,3-dihydro-1,4-benzodioxin-6-yl-5-5-nitro-1,3-thiazol-2-yl sulfanyl-2h-1,2,4-triazol-3-one, bi-78d hplc, 4-2,3-dihydro-1,4-benzodioxin-6-yl-3-5-nitro-1,3-thiazol-2-yl sulfanyl-4,5-dihydro-1h-1,2,4-triazol-5-one, 4-2,3-dihydro-1,4-benzodioxin-6-yl-3-5-nitro-2-thiazolyl thio-1h-1,2,4-triazol-5-one, 4-2,3-dihydro-1,4-benzodioxin-6-yl-5-5-nitro-1,3-thiazol-2-yl sulfanyl-1,2,4-triazol-3-ol, 4-2,3-dihydro-1,4-benzodioxin-6-yl-5-5-nitro-1,3-thiazol-2-yl sulfanyl-4h-1,2,4-triazol-3-ol
PubChem CID 2747117
IUPAC Name 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1H-1,2,4-triazol-5-one
SMILES C1COC2=C(O1)C=CC(=C2)N3C(=O)NN=C3SC4=NC=C(S4)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 379.37
Percent Purity >99%
Chemical Name or Material BI 78D3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.