Learn More
Bicinchoninic acid disodium salt, Thermo Scientific Chemicals
Description
For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 2 g |
Solubility Information | Soluble in water. |
IUPAC Name | disodium [2,2'-biquinoline]-4,4'-dicarboxylate |
Formula Weight | 388.29 |
Physical Form | Chunks and/or Powder |
Chemical Name or Material | Bicinchoninic acid disodium salt |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U07G017 | Certificate of Analysis | 15/01/2025 | 044247.04,044247.09 |
X22F059 | Certificate of Analysis | 14/05/2024 | 044247.04,044247.09 |
Q08K029 | Certificate of Analysis | 12/04/2024 | 044247.04,044247.09 |
Y01J003 | Certificate of Analysis | 05/12/2023 | 044247.04,044247.09 |
Y29I020 | Certificate of Analysis | 25/01/2023 | 044247.04,044247.09 |
Safety and Handling

- Bicinchoninic acid, disodium salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Bicinchoninic acid,disodium salt
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.