missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 99+%, for analysis, Thermo Scientific Chemicals

100.40 € - 476.37 €

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10428320
View Documents
Thermo Scientific Acros
172651000
100 g Glass bottle
100.40 €
100g
Please to purchase this item. Need a web account? Register with us today!
10742564
View Documents
Thermo Scientific Acros
172652500
250 g Glass bottle
194.97 €
250g
Please to purchase this item. Need a web account? Register with us today!
10667943
View Documents
Thermo Scientific Acros
172650010
1 kg Glass bottle
476.37 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

150-25-4
185.16
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
8761
2-[bis(2-hydroxyethyl)amino]acetic acid
C6H12NNaO4
MFCD00004295
N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
CHEBI:40957
[Na+].OCCN(CCO)CC([O-])=O

Specifications

0.05 max. (5% aqueous soln.) at 250nm
150-25-4
100.0
4.0 to 5.5 (10% soln. at 20°C)
99+%
C6H12NNaO4
MFCD00004295
N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
[Na+].OCCN(CCO)CC([O-])=O
185.16
CHEBI:40957
≥99.0% (Total base)
Bicine, 99+%
(5% aq. soln.) (1cm cell)
99.0
White
Crystalline Powder
Authentic
(HOCH2CH2)2NCH2CO2H
15, 1205
MFBDBXAVPLFMNJ-UHFFFAOYSA-M
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
163.17
Analytical
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-755-1

missing translation for 'rtecsNumber' : MB9700000

missing translation for 'tsca' : TSCA

Product Identifier
  • Bicine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.