missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 97%, Thermo Scientific Chemicals

53.37 € - 141.71 €

Chemical Identifiers

CAS 1719-83-1
Molecular Formula C12H8O6
Molecular Weight (g/mol) 248.19
MDL Number MFCD00082228
InChI Key XLOGCGOPKPCECW-UHFFFAOYSA-N
Synonym bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
PubChem CID 102679
SMILES C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315279
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A19632.14
25 g
53.37 €
25g
Please to purchase this item. Need a web account? Register with us today!
11325279
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A19632.22
100 g
141.71 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride is used to prepare the thermo-cured cycloaliphatic polyamic acid and photo-cured cycloaliphatic polyamic ester, which were then converted to cycloaliphatic polyimide film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride is used to prepare the thermo-cured cycloaliphatic polyamic acid and photo-cured cycloaliphatic polyamic ester, which were then converted to cycloaliphatic polyimide film.

Solubility
Soluble in water

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

1719-83-1
248.19
XLOGCGOPKPCECW-UHFFFAOYSA-N
102679
C12H8O6
MFCD00082228
bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O

Specifications

1719-83-1
C12H8O6
294213
bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone, bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride, 4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride, bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride, 4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone, 4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone, bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride, 4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
XLOGCGOPKPCECW-UHFFFAOYSA-N
248.19
248.19
Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride
∼360°C
MFCD00082228
Moisture sensitive
Soluble in water
C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O
102679
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 217-009-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only