Learn More
Bifenazate, MedChemExpress
MedChemExpress Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm. Bifenazate is a positive allosteric modulator of GABA receptor.
Brand:  Medchem Express HY-119687
Encompass_Preferred
 View more versions of this product
Product Code. 16431219
  • 159.30 € / 1g
Explore more special offers
Description
Description
Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm. Bifenazate is a positive allosteric modulator of GABA receptor.
Specifications
Specifications
Bifenazate
Yellow
C17H20N2O3
DMSO : ≥ 250 mg/mL (832.36 mM)
300.35
99.65%
Solid
149877-41-8
Research
1 g
COC1=CC=C(C2=CC=CC=C2)C=C1NNC(OC(C)C)=O
300.35
Research
Neuroscience-Neuromodulation