missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ Big-Digit Stopwatch with Calibration
The jumbo digits and high accuracy of the Traceable Big-Digit Stopwatch with Calibration make it the perfect stopwatch for the lab or field. Display time of day, date and day of week. The ABS plastic case is water-, dust- and shock-resistant, with a handy clip and hoop and loop fasteners allows you to mount the stopwatch to various surfaces.
Brand: Traceable™ 94460-28
31.61 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Nist traceable
- Easy to read jumbo digits
- High accuracy
- Includes time of day alarm
- Water-, dust- and shock-resistant
- Mounts to various surfaces due to its handy clip, and hoop and loop fastener
- Single-action, time-out, cumulative-split and continuous timing functions
- Neck cord
Specifications
24 hr. | |
±0.001% | |
Big-Digit Stopwatch | |
1.5 V Button Cell | |
Neck Cord |
Audible | |
0.01 sec, 1 sec | |
ABS Plastic | |
5/8 in. | |
NIST Traceable Certificat Supplied, ISO/IEC 17025:2017(1750.01) Calibration Laboratory Accredited by A2LA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Traceable™ Big-Digit Stopwatch with Calibration > Stopwatch with Calibration; Time Range: 24 hours; Battery Type: 1.5 V Button Cell; Resolution: 0.01 sec, 1 sec; Alarm: Audible, 1 min. Beep; Material: ABS Plastic;
Spot an opportunity for improvement?Share a Content Correction