Learn More
Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.
1848.00 €
Specifications
Format | 12-well Strips, 8-well Strip, 96- or 384-well Plate |
---|---|
For Use With (Equipment) | SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers |
Product Line | BigDye™ Terminator |
Product Type | Sequencing Buffer |
Technique | Fluorescent Dye Terminator Sequencing |
Description
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.
- Improve the quality of your results for a wide range of sequencing applications
- Optimized for long read lengths
- Better dye mobility characteristics
- Improved performance reading through GT-rich regions
- Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
- Enhance your productivity and reduce costs
Get more uniform peak heights and improved accuracy
With BigDye™ Terminator v3.1 kit superior chemistry, you generate data with uniform peak heights and optimized signal balance. This results in longer, higher-quality reads and more accurate base assignments for heterozygote and mutation detection.
Specifications
12-well Strips, 8-well Strip, 96- or 384-well Plate | |
BigDye™ Terminator | |
Fluorescent Dye Terminator Sequencing |
SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers | |
Sequencing Buffer |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.