Learn More
Invitrogen™ Bigfoot™ Spectral Cell Sorter Accessories
The Bigfoot Cell Sorter is designed to streamline the cell sorting workflow and enhance the user experience. It has a multitude of cell sorting advancements, accessories, and unique capabilities to enable the most complex multi-parameter sorts, and to meet high-throughput demands for a wide range of applications.
Brand: Invitrogen™ PL00355
Description
The Invitrogen Bigfoot Spectral Cell Sorter provides power, performance, and flexibility to help accelerate cutting edge discoveries. Designed to streamline the cell sorting workflow and enhance the user experience, the Bigfoot sorter has a multitude of cell sorting advancements, accessories, and unique capabilities to enable the most complex multi-parameter sorts, and to meet high-throughput demands for a wide range of applications.
The Invitrogen Bigfoot Spectral Cell Sorter offers a full line of accessory products for continuous expansion of the instrument including interchangeable nozzle tips, temperature controlled sample input and sort output adapters, and bulk fluid bottles and caps.Specifications
1.5 mL | |
Flow Cytometry | |
Room Temperature | |
Bigfoot™ |
Bigfoot™ Spectral Cell Sorter | |
Each | |
Tube Adapter | |
Output Tube Adapter, 1.5 mL |