Learn More
Thermo Scientific™ Bile Esculin Agar
Isolate and differentiate enterococci / Group D streptococci and non-Group D streptococci using this selective microbiological medium.
21.60 €
Product Code | Brand | Description | Format | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Format | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
12355743
|
Thermo Scientific™
|
Bile Esculin Agar | 90mm Monoplate | 10/Pk. | United Kingdom |
21.60 €
Each |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
Description
Isolate and differentiate enterococci/Group D streptococci and non Group D streptococci using Thermo Scientific™ Bile Esculin Agar, a selective microbiological medium.
Enterococci / Group D streptococci hydrolyse aesculin to form aesculetin and dextrose. Aesculetin combines with ferric citrate in the medium to form a dark brown or black complex which is indicative of a positive result. Bile salts will inhibit Gram-positive bacteria other than enterococci / Group D streptococci.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Bile Esculin Agar | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 9 Weeks