missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ BIO-SILICO™ Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Specially processed culture plugs with coarse bubbles for preparation, filling and sterilization of culture media.
Specifications
Color | Orange |
---|---|
Material | Silicone |
Product Type | Silicone Stopper |
For Use With (Application) | For The Preparation And Sterilization Of Culture Media |
Quantity | 10/Pack |
Description
- Coarse pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Orange | |
Silicone Stopper | |
10/Pack |
Silicone | |
For The Preparation And Sterilization Of Culture Media |