Learn More
Thermo Scientific™ BioBasic™ PREP SEC 120 Size Exclusion Chromatography HPLC Columns
Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 120 Size Exclusion Chromatography HPLC Columns.
816.09 € - 2527.12 €
Specifications
Diameter (Metric) | 7.8 mm |
---|---|
For Use With | Preparative Size Exclusion |
Max. Pressure | 4300 psi (295 bar) |
Particle Size | 5 μm |
pH | 2 to 8 |
Product Code | Brand | Length (Metric) | Column Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Column Format | Price | Quantity & Availability | |||||
16651674
|
Thermo Scientific™
73405-037821A |
30 mm | Preparative Guard Column |
816.09 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
16661674
|
Thermo Scientific™
73405-157846A |
150 mm | Preparative Column, Standard |
1690.59 €
Each |
In Stock
Log in to see stock. |
|||||
16515944
|
Thermo Scientific™
73405-307846A |
300 mm | Preparative Column, Standard |
2527.12 €
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
|||||
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 700 to 120,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.
BioBasic™ PREP SEC 120 HPLC Column Benefits:
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 120Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
7.8 mm | |
4300 psi (295 bar) | |
2 to 8 | |
300 m²/g | |
L59 | |
Spherical, Fully Porous, Ultrapure Silica | |
Size Exclusion | |
Size Exclusion |
Preparative Size Exclusion | |
5 μm | |
120 Å | |
60°C | |
Diol (OH) | |
>8.2% | |
No | |
Preparative Chromatography |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.