missing translation for 'onlineSavingsMsg'
Learn More
Ansell BioClean™ Beard Snood BBS-W Product Code.: 15482480
Add to basket

Ansell BioClean™ Beard Snood BBS-W

Product Code. 15482480
Color: White
Click to view available options
Color: White
This item is not returnable. View return policy

Product Code. 15482480

Brand: Ansell BBSW

Add to basket
Add to basket
This item is not returnable. View return policy

Ansell BioClean™ Beard Snood BBS-W is manufactured from lightweight, soft, hupoallergenic nonwoven material.

  • Lightweight
  • Comfortable headloop
  • Latex-free elastic opening
  • Universal size
TRUSTED_SUSTAINABILITY

Specifications

Type Cleanroom Beard Cover
Size Universal
Material Spunbond Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Seam Type Double Stitched Opening
Certifications/Compliance ISO 9001:2015
For Use With (Application) Cleanrooms
Packaging Quantity 100 pieces per sealed inner PE bag; 10 inner bags per sealed outer PE bag; one outer bag per lined carton (1000 pieces)
Weight (Metric) 106 g (±2 g)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell BioClean™ Beard Snood BBS-W >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.