Learn More
Nitritex™ BioClean Permaflow™ Sterile Cleanroom Marker Pens
Processed to ensure ISO Class 4 & EU GMP Grade A compatibility. Nitritex™ BioClean Permaflow™ Cleanroom Marker Pens are individually packaged and available with black, blue, or red ink. Double-bagged varieties are available.
Brand:  Nitritex™ S-BPFP-BK1
22.13 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15994841
  • 31.85 € / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Super permanent ink in black, blue, or red
  • Fade and water resistant
  • Quick drying
  • Non-toxic
  • Alcohol resistant labeling
  • Autoseal ink prevents drying
  • Store in a dry, cool place away from direct sunlight and fluorescent lighting
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Processed in an NEBB certified ISO Class 4 cleanroom environment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Black
Double bagged
5.5 in.
140 mm