Learn More
Teknon™ Biocleanse Ready to Use
Use in scientific, medical and biotechnology environments to combat biological hazards.Teknon™ Biocleanse Ready to Use is a convenient deodorising cleaner, degreaser, disinfectant and fungicide.
Brand: Teknon™ D/0007/21
Description
Use in scientific, medical and biotechnology environments to combat biological hazards.Teknon™ Biocleanse Ready to Use is a convenient deodorising cleaner, degreaser, disinfectant and fungicide.
Ready to use is equivalent to a 5% solution of Biocleanse Concentrate. This product has powerful bactericidal and virucidal properties. Biocleanse is free from sodium hypochlorite, phosphates and enzymes and effectively removes blood, fat, proteins, grease and other organic and non-organic contaminants whilst simultaneously disinfecting the treated surface. It is safe to use on ferrous and non-ferrous metals, ceramics, glassware and plastics.
Biocleanse meets the requirements of BS EN 1276 and is effective against MRSA, HIV, HBV, Avian Flu, E coli, Salmonella, Listeria, Candida, Penicillium and C. diff.
Under COSHH Regulations, the product does not require a maximum exposure limit and is not irritating to skin.
- Wide spectrum biocide
- Effective against C. diff, MRSA, HIV, HBV and Avian Flu
- Passes BS EN 1276
- Powerful cleaner
- Ready to use, non tainting
Specifications
Liquid | |
Bulk Cannister | |
5 L |
Blue | |
For use in scientific, medical and biotechnology environments to combat biological hazards |
Safety and Handling
- 5 LT BIOCLEANSE READY TO USE BIOCIDAL CLEANER 5L
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P262-Do not get in eyes, on skin, or on clothing.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : Didecyldimethyl ammonium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.