Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
579.94 € - 2663.09 €
Specifications
Pore Size | 3 μm |
---|---|
Type | Membrane |
For Use With (Application) | Endothelial Cell Invasion |
No. of Wells | 24 |
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifications
3 μm | |
Membrane | |
Endothelial Cell Invasion | |
24 |