Learn More
Corning™ BioCoat™ Endothelial Cell Growth Environment Kit
Increases endothelial cell yield from a variety of sources
434.30 € - 479.36 €
Specifications
For Use With (Application) | Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology |
---|---|
Storage Requirements | 2° to 8°C |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10365771
|
Corning™
355053 |
Complete Endothelial Cell Growth Environment Kit | |||||||
11523590
|
Corning™
355054 |
Endothelial Cell Culture Medium |
434.30 €
1 set |
Estimated Shipment: 31-01-2025
Log in to see stock. |
|||||
Description
Ideal for human umbilical vein (HUVEC), pulmonary artery (HPAEC), and aorta (HAEC), as well as fetal bovine heart (FBHEC) cells. Interactions between components of the extracellular matrix and growth factors play a crucial role in determining the adaptation and responses of mammalian cells to their micro environments both in vivo and in vitro.
- Cells can be subcultured according to routine procedures, seeded into fermentation systems, or used in a variety of bioassays
- Integrate specially formulated low-serum or serum-free medium, culture supplements and growth factors
- Each environment lot is tested in a bioassay and all components are tested for bacteria and fungi
These products are for research use only and are not intended for diagnostic purposes.
Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology
Order Info
Media and cellware are also available separately.
Specifications
Construction of model systems to study angiogenesis, atherosclerosis, wound healing and in vitro toxicology | |
2° to 8°C |