missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap
Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells
321.00 € - 496.44 €
Specifications
Closure Material | Phenolic |
---|---|
For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
Neck Style | Canted |
Shape | Rectangular |
Surface Coating | Laminin |
Product Code | Brand | Capacity (Metric) | Culture Area | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Culture Area | Price | Quantity & Availability | ||||
10536381
|
Corning™
354522 |
250 mL | 75 cm2 |
496.44 €
Pack of 10 |
Estimated Shipment: 27-03-2025
Log in to see stock. |
||||
10719241
|
Corning™
354533 |
70 mL | 25 cm2 | N/A | |||||
Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
These products are for research use only and are not intended for diagnostic purposes.
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Phenolic | |
Canted | |
Laminin |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Rectangular | |
Sterile |