missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Biocontainment Lid for Thermo Scientific™ Sorvall™ Microplate Rotor 75-002-010
Optional biocontainment-approved lids offered in pairs
Brand: Thermo Scientific™ 75002011
237.74 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Accessory for Sorvall™ MP 3300 Microplate Rotor
For use with Sorvall microplate rotor 75-002-010. Sold in pairs.
Specifications
Biocontainment Lid | |
75-002-010 Microplate rotor | |
MP 3300 Microplate Rotor accessory |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction