missing translation for 'onlineSavingsMsg'
Learn More

Biocytin, Tocris Bioscience™

Product Code. 18094732
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Versatile marker for neuroanatomical investigations

Versatile marker used in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays. Displays high solubility in aqueous solutions and has a low molecular weight facilitating injection using micropipettes. Can be incorporated with a variety of avidin and streptavidin conjugates for detection by light, fluorescence or electron microscope.

Chemical Identifiers

CAS 576-19-2
Molecular Formula C16H28N4O4S
Molecular Weight (g/mol) 372.484
InChI Key BAQMYDQNMFBZNA-MNXVOIDGSA-N
Synonym biocytin, n'-biotinyl-l-lysine, biotinyl-l-lysine, h-lys biotinyl-oh, +-biocytin, ne-biotynyl-l-lysine, epsilon-n-biotinyl-l-lysine, n6-d-biotinyl-l-lysine, n6-5-3as,4s,6ar-hexahydro-2-oxo-1h-thieno 3,4-d imidazol-4-yl-1-oxopentyl-l-lysine, n6-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoyl-l-lysine
PubChem CID 83814
ChEBI CHEBI:27870
IUPAC Name (2S)-6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-2-aminohexanoic acid
SMILES C1C2C(C(S1)CCCCC(=O)NCCCCC(C(=O)O)N)NC(=O)N2

Specifications

Quantity 50 mg
Formula Weight 372.48
Percent Purity >98%
Chemical Name or Material Biocytin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.