missing translation for 'onlineSavingsMsg'
Learn More

Biofuel Standard, SPEX CertiPrep™

Product Code. 11945084
Click to view available options
Quantity:
100 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 67784-80-9, 7704-34-9
Packaging Triple-leached LDPE bottle
Quantity 100 mL
CAS Max % 99.99, 0.01
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition (by Analyte or Components) 100μg/g
Analyte or Component Names Biodiesel B-100,Sulfur from Ammonium sulfate
Solvent or Matrix B100
Chemical Name or Material Biofuel Standard, For Biodiesel Fuel Analysis

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'