missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific™ Biohazard Disposal Bags
Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.
Brand: Heathrow Scientific™ HS10320
72.71 EUR valid until 2024-10-31
Use promo code "21744" to get your promotional price.
This item is not returnable.
View return policy
Description
- Biohazard collection bag
- Biohazard warning symbol and precautions printed in English, Spanish, and French
- Includes an indicator patch that darkens when autoclaved
- Autoclavable to 250°F (121°C)
- Disposable
- HS10044H bag holder is ONLY for use with HS10320 2 L bag
Specifications
Benchtop Biohazard Disposal Bag | |
Autoclavable | |
2 L | |
Single-use | |
Autoclave | |
Non-sterile | |
500 Pk. | |
63 mil | |
8 x 12 x 0.06 in. | |
20.3 cm | |
30.5 cm | |
0.15 cm | |
0.01 kg |
Biohazard Disposal Bags | |
0.5 gal. | |
Red | |
Biohazard Disposal | |
Polypropylene | |
1.6 mm | |
Box | |
8 x 12 in. | |
8 in. | |
12 in. | |
0.06 in. | |
0.01 lb. |