missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Disposal Bags with or without Warning Label

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes

Brand:  Bel-Art™ F13161-0005

 View more versions of this product

Product Code. 11302154

  • 146.01 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes

  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners
Specifications

Specifications

Biohazard Bag
12 gal.
ASTM D1709-98
Translucent
Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Polypropylene
1.5 mm
30 x 24 in. (76 x 61 cm)
76 cm
61 cm
Autoclavable
45 L
Zip Tie
Single-use
Non-printed without Sterilization Patch
Non-sterile
1.5 mil
30 in.
24 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.