Learn More
Bel-Art™ Biohazard Disposal Bags with or without Warning Label
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
Brand: Bel-Art™ F13162-0005
164.08 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Description
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
- These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
- Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
- High temperature autoclavable at a maximum of 135°C (275°F)
- May also be used as conventional trash can liners
Specifications
Biohazard Bag | |
20 gal. | |
ASTM D1709-98 | |
Translucent | |
Disposal of used pipettes, petri dishes, culture plates, membrane filters and more | |
Polypropylene | |
1.5 mm | |
36 x 24 in. (91 x 61 cm) | |
91 cm | |
61 cm |
Autoclavable | |
170 L | |
Zip Tie | |
Single-use | |
Non-printed without Sterilization Patch | |
Non-sterile | |
1.5 mil | |
36 in. | |
24 in. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.