Learn More
IBA Lifesciences BioLock
GREENER_CHOICE
BioLock is a Biotin blocking solution for lysates and culture supernatants
240.00 € - 959.00 €
Specifications
Type Blocking Solution
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15643608
GREENER_CHOICE
View Documents
IBA Lifesciences
2-0205-050
50 mL
240.00 €
50mL
In Stock
to see stock.
Edge
Add to basket
15653608
GREENER_CHOICE
View Documents
IBA Lifesciences
2-0205-250
250 mL
959.00 €
250mL
Estimated Shipment: 22-01-2025
to see stock.
Add to basket
Description
Description
BioLock is a ready-to-use biotin blocking solution which contains the tetrameric biotin binding protein avidin. BioLock prevents biotin and biotinylated proteins from binding to Strep-Tactin™ or Strep-Tactin™XT, whereas binding of the Twin-Strep-tag™ and Strep-tag™II is not influenced.
Biotin and/or biotinylated proteins can possibly reduce the binding capacity of Strep-Tactin™ as well as Strep-Tactin™XT and are present in cell lysates and cell culture media, especially from mammalian or insect cells. Thus, if Twin-Strep-tag™ and Strep-tag™II fusion proteins from biotin-containing extracts or media are intended to be purified via Strep-Tactin™ or Strep-Tactin™XT affinity chromatography, biotin has to be masked by addition of BioLock prior to application onto the resin. 
Specifications
Specifications
Blocking Solution