missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ BioNick™ DNA Labeling System
BioNick™ DNA Labeling System
Brand: Molecular Probes™ 18247015
405.35 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The BioNick™ DNA Labeling System is ideal for generating biotinylated DNA probes by nick translation optimized for use in non-radioactive in situ hybridizations. These probes can also be used in Southern or northern blots, plaque lifts, colony hybridizations, and dot blot hybridizations. Using the BioNick™ DNA Labeling System:Specifications
The BioNick™ DNA Labeling System includes 10X dNTP mix containing biotin-14-dATP, 10X enzyme mix containing DNA Polymerase I and DNase I, control DNA, stop buffer, and distilled water. Store at -20°C. Guaranteed stable for 6 months when properly stored. | |
Biotin | |
DNA (General) | |
Direct Labeling | |
DNA Labeling System | |
Approved for shipment on Wet or Dry Ice |
Biotin-based | |
Probes (Labeled DNA) | |
Yes | |
BioNick™ | |
50 Reactions |
For Research Use Only. Not for use in diagnostic procedures.