Learn More
Simport™ Scientific Biopsy Foam Pads
Used to hold biopsies in place and prevent them from being lost during processing.
Description
Description
  • Made of polyester urethane specially formulated foam for throughout consistency to achieve optimum solvent flow
  • Biopsy samples are sandwiched between two foam pads and are placed either in tissue capsules or cassettes with metal or plastic lids
  • Resist temperatures from −40°C to +121°C