Learn More
Unity Lab Services™ Biosafety Cabinet IQOQ Service
IQOQ service for class II biosafety cabinets.
Brand:  Unity Lab Services™ IOQBSCMVS
Product Code. 17927792
  • 1890.00 € / 1 unit
Explore more special offers
Description
Description
  • IQ:Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order.
  • Verifies equipment and any other accessories are assembled and installed. Any abnormal event(s) observed during the IQ is documented in the IQ protocol.
  • Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements.
  • Configuration information for each system component gets recorded.
  • Installation of Equipment is NOT Included.
  • OQ: System component information detailed configuration information for each system component have been recorded (as applicable).
  • Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
  • Key test varies based on relevant certification standard (NSF49/EN12469/DIN12480). Examples: filter leakage, DEHS, air velocity, electrical safety, air-flow smoke pattern, KI-DISCUS™.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biosafety Cabinets
IQ/OQ Service
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products